2-Methyl-1-pentene, Supelco®
Catalog # 68450-25ML
Supplier: Merck
CAS Number:
Specifications
- Pack type:Glass bottle
- Pk:25 mL
- Seq:2315
Specifications
About this item
:
:84.16 g/mol
Boiling Pt:62 °C (1013 hPa)
Melting Pt:–135.76 °C
Density:0.7083 g/cm³ (0 °C)
Storage Temperature:Ambient
EINECS:212-108-7
UN:2288
ADR:3,II
:M-Clarity™ quality level = MQ100
CAS Number:763-29-1
Formula:C₆H₁₂
MDL Number:MFCD00009405