


2-Methyl-1-pentene 99%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:763-29-1
:
Formula:C₆H₁₂
MDL Number:MFCD00009405
:84.16 g/mol
Boiling Pt:62 °C (1013 hPa)
Melting Pt:–136 °C
Density:0.68 g/cm³
Flash Pt:–26 °C
Storage Temperature:Refrigerator
EINECS:212-108-7
UN:3295
ADR:3,II
:GHS07
:GHS08
:GHS02
Specification Test Results
- Appearance:Clear colorless Liquid
- Infrared spectrum:Conforms
- Refractive index:1.3910 to 1.3930 (20°C, 589 nm)
- GC:≥98.5 %