
1-Hexadecene for synthesis, Sigma-Aldrich®
Supplier: Merck
Certificates
About this item
CAS Number:629-73-2
:
Formula:C₁₆H₃₂
MDL Number:MFCD00008991
:224.43 g/mol
Boiling Pt:285 °C (1013 hPa)
Melting Pt:4 °C
Density:0.7811 g/cm³ (20 °C)
:M-Clarity™ quality level = MQ200
Flash Pt:128 °C
EINECS:211-105-8
:GHS08
Storage Temperature:Ambient
Specification Test Results
- Solubility:0.0012 g/l (25 °C)
- Melting Point:2.2 °C
- Molar Mass:224.42 g/mol
- Boiling Point:274 °C (1013 hPa)
- Vapor Pressure:0.35 hPa (25 °C)
- Flash Point:132 °C
- Refractive Index:1.4401 (20 °C, 589 nm)
- Explosion Limit:0.5 - 5.8 %(V)
- Density:0.78 g/cm3 (20 °C)
- Ignition Point:240 °C
- Viscosity Kinematic:4.0 mm2/s (20 °C)