Order Entry
Finland
ContactUsLinkComponent
1-Hexadecene 92% tech.
Health
1-Hexadecene 92% tech.
Catalog # ACRO120500010
CAS Number:  
undefined
Health
1-Hexadecene 92% tech.
Catalog # ACRO120500010
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    1 L
  • Seq:
    5037

Specifications

Specification Test Results

  • Appearance:
    Clear colorless Liquid
  • Infrared spectrum:
    Conforms
  • Refractive index:
    1.4390 to 1.4430 (20°C, 589 nm)
  • GC:
    ≥90 %

Specification Test Results

About this item

 :
 :224.43 g/mol
Boiling Pt:285 °C (1013 hPa)
Melting Pt:3…5 °C
Density:0.783 g/cm³
Flash Pt:132 °C
Storage Temperature:Ambient
EINECS:211-105-8
 :GHS08
CAS Number:629-73-2
Formula:C₁₆H₃₂
MDL Number:MFCD00008991