

trans-2-Pentenal 97%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:1576-87-0
:
Formula:C₅H₈O
MDL Number:MFCD00009615
:84.12 g/mol
Boiling Pt:127 °C (1013 hPa)
Density:0.86 g/cm³
Flash Pt:19 °C
Storage Temperature:Refrigerator
EINECS:216-414-1
UN:1993
ADR:3,II
:GHS07
:GHS02
Specification Test Results
- Appearance:Clear colorless to yellow Liquid
- Infrared spectrum:Conforms
- Refractive index:1.4400 to 1.4470 (20°C, 589 nm)
- GC:≥96.0 %