Benzyl acetate ≥99%
About this item
Purity: 99% CAS No.: 140-11-4 Molecular Formula: CH3CO2CH2C6H5 Formula Weight: 150.18 Flash Point: 95°(203°F) Physical Form: Liquid Density: 1.053 Refractive Index: 1.5020 Appearance: Colourless Odour: Fruit-like Melting Point: -51° Boiling Point: 205-207° Beilstein Reference: 1908121 Merck Reference: 14,1123 MDL No.: MFCD00008712
- CAS Number:140-11-4
- Formula:C₉H₁₀O₂
- MDL Number:MFCD00008712
- Molecular Weight:150.18 g/mol
- Merck Index:14,1123
- Boiling Pt:205…207 °C
- Melting Pt:–51 °C
- Density:1.053
- Flash Pt:95 °C (203 °F)
- Storage Temperature:Ambient
- EINECS:205-399-7
2 Options Available Below
Product Details & Documents
Purity: 99% CAS No.: 140-11-4 Molecular Formula: CH3CO2CH2C6H5 Formula Weight: 150.18 Flash Point: 95°(203°F) Physical Form: Liquid Density: 1.053 Refractive Index: 1.5020 Appearance: Colourless Odour: Fruit-like Melting Point: -51° Boiling Point: 205-207° Beilstein Reference: 1908121 Merck Reference: 14,1123 MDL No.: MFCD00008712
- CAS Number:140-11-4
- Formula:C₉H₁₀O₂
- MDL Number:MFCD00008712
- Molecular Weight:150.18 g/mol
- Merck Index:14,1123
- Boiling Pt:205…207 °C
- Melting Pt:–51 °C
- Density:1.053
- Flash Pt:95 °C (203 °F)
- Storage Temperature:Ambient
- EINECS:205-399-7
Documents
Product Family Options
Product Information
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



