Screw couplings for threaded tubes
Supplier: Lenz Laborglas GmbH & CO.KG
Certificates
About this item
Connectors, made of PBT, red with integrated silicone (VMQ)-PTFE sealing.
- For flexible connection of two DIN glass threads
- Heat resistant from –45 to +180 °C
- Extremely leak tight