Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Finland
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Stirrers
Magnetic Stirring Bars
Magnetic stirring bars, PTFE
Magnetic stirring bars, PTFE
Magnetic stirring bars, PTFE
Supplier:
IKA
Certificates
Expand image
Magnetic stirring bars, PTFE
Supplier:
IKA
About this item
Very good chemical resistance
Temperature resistant, suitable for high and low temperature applications