Septum caps, Nalgene®
38 - 430, PP
Supplier: Thermo Fisher Scientific
Septum caps, Nalgene®
Catalog # 391-0073
Supplier: Thermo Fisher Scientific
Jump to:
Product Details & Documents
Polypropylene closure with thermoplastic elastomer septum.
- Use with 18-gauge or smaller needles
For Nalgene bottles with 38-430 threaded mouth.
Documents
Certificates
Search by Lot NumberSpecifications
- Cap Size:38 - 430
- Material:PP
- Cat. No.:391-0073
- Environmental certification:NAR Certificate of Retirement
- Environmentally Preferable:Low Manufacturing Impact
Suositukset