Variable dispenser with bottle, Nalgene®
500 ml, White/clear
Catalog # NALG3702-0025
Supplier: Thermo Fisher Scientific
Jump to:
Product Details & Documents
Variable-volume bottle dispenser, supplied with a LD-PE bottle and fill tube.
- PMP measuring chamber
- PP stem closure and push-pull stopper
- HD-PE ring-tab
The dispenser is leakproof when patented push-pull stopper on end of stem is closed. Ring-tab fits snugly around bottle neck for attaching a label. Designed to dispense bases, aqueous solutions and alcohols.
Specifications
- Volume:500 ml
- Colour:White/clear