n-Hexadecane, Supelco®
Supplier: Merck
Jump to:
About this item
Synonyms: Hexadecane, Cetane
- CAS Number:544-76-3
- Formula:H₃C(CH₂)₁₄CH₃
- MDL Number:MFCD00008998
- Molecular Weight:226.45 g/mol
- Boiling Pt:287 °C (1013 hPa)
- Melting Pt:18 °C
- Density:0.77331 g/cm³ (20 °C)
- :M-Clarity™ quality level = MQ100
- Flash Pt:135 °C (closed cup)
- Storage Temperature:Ambient
- EINECS:208-878-9
2 Options Available Below
Product Details & Documents
Synonyms: Hexadecane, Cetane
- CAS Number:544-76-3
- Formula:H₃C(CH₂)₁₄CH₃
- MDL Number:MFCD00008998
- Molecular Weight:226.45 g/mol
- Boiling Pt:287 °C (1013 hPa)
- Melting Pt:18 °C
- Density:0.77331 g/cm³ (20 °C)
- :M-Clarity™ quality level = MQ100
- Flash Pt:135 °C (closed cup)
- Storage Temperature:Ambient
- EINECS:208-878-9



