Order Entry
Finland
ContactUsLinkComponent
Glycerine ≥97%, TECHNICAL
Glycerine ≥97%, TECHNICAL
Supplier:  VWR Chemicals
   Sustainable Options Available
Certificates
undefined
Glycerine ≥97%, TECHNICAL
Supplier:  VWR Chemicals
   Sustainable Options Available
Certificates

Specifications

  • Environmentally Preferable:
    Safer Human and Environmental Health
  • Environmental certification:
    EPA Safer Chemical Solvent (Green Circle Rating)

Specifications

Specification Test Results

  • Assay:
    Min. 97 %

Specification Test Results