Order Entry
Finland
ContactUsLinkComponent
Glycerine ≥99.5%, AnalaR NORMAPUR® ACS analytical reagent, redistilled
Glycerine ≥99.5%, AnalaR NORMAPUR® ACS analytical reagent, redistilled
Supplier:  VWR Chemicals
   Sustainable Options Available
Certificates
undefined
Glycerine ≥99.5%, AnalaR NORMAPUR® ACS analytical reagent, redistilled
Supplier:  VWR Chemicals
   Sustainable Options Available
Certificates

Specifications

  • Environmentally Preferable:
    Safer Human and Environmental Health
  • Environmental certification:
    EPA Safer Chemical Solvent (Green Circle Rating)

Specifications

Specification Test Results

  • Assay:
    Min. 99.5 %
  • Acrolein and glucose:
    Passes test
  • Appearance of solution (10 % V/V; water):
    Passes test
  • Substances darkened by sulphuric acid:
    Passes test
  • Acidity:
    Max. 0.0004 meq/g
  • Alkalinity:
    Max. 0.0003 meq/g
  • Colouration:
    Max. 10 APHA
  • Density (20/4):
    1.257 - 1.265
  • Acetaldehyde:
    Max. 10 ppm
  • Esters of fatty acids (as C15H26O6):
    Max. 0.05 %
  • Glyceraldehyde:
    Max. 30 ppm
  • Heavy metals (as Pb):
    Max. 1 ppm
  • Ignition residue (SO4):
    Max. 50 ppm
  • Ketones (as CH3COCH3):
    Max. 50 ppm
  • Water:
    Max. 0.5 %
  • Cl (Chloride):
    Max. 1 ppm
  • NH4 (Ammonium):
    Max. 5 ppm
  • SO4 (Sulphate):
    Max. 5 ppm
  • As (Arsenic):
    Max. 1 ppm
  • Ba (Barium):
    Max. 5 ppm
  • Fe (Iron):
    Max. 1 ppm
  • Conforms to ACS:
    Passes test

Specification Test Results