Order Entry
Finland
ContactUsLinkComponent
Glycerine ≥99.5% for fluorescence microscopy, Sigma-Aldrich®
Glycerine ≥99.5% for fluorescence microscopy, Sigma-Aldrich®
Catalog # 1.04095.0250
Supplier:  Merck
CAS Number:  
undefined
Glycerine ≥99.5% for fluorescence microscopy, Sigma-Aldrich®
Catalog # 1.04095.0250
Supplier:  Merck
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    250 ml
  • Seq:
    2590
  • Pk Info:
    Preparation Capacity 1250 slides

Specifications

Specification Test Results

  • Purity (GC):
    ≥ 99.5 %
  • Identity (IR-spectrum):
    passes test
  • Refractive index (n 20/D):
    1.472 - 1.474
  • Fluorescence (as quinine at 365 nm):
    ≤ 100 ppb
  • Fluorescence (as quinine at 313 nm):
    ≤ 100 ppb
  • Fluorescence (as quinine at 254 nm):
    ≤ 100 ppb
  • Transmission (at 240 nm; 1 cm):
    ≥ 60 %
  • Transmission (at 260 nm; 1 cm):
    ≥ 75 %
  • Transmission (at 280 nm; 1 cm):
    ≥ 80 %
  • Transmission (from 300 nm; 1 cm):
    ≥ 85 %
  • Suitability for fluorescence microscopy:
    passes test

Specification Test Results

About this item

 :
 :92.09 g/mol
Merck Index:13,04497
Boiling Pt:290 °C (1013 hPa)
Melting Pt:18.6 °C
Density:1.26 g/cm³ (20 °C)
 :M-Clarity™ quality level = MQ300
Flash Pt:199 °C
Storage Temperature:Ambient
EINECS:200-289-5
 :1
CAS Number:56-81-5
Formula:HOCH₂CH(OH)CH₂OH
MDL Number:MFCD00004722