Order Entry
Finland
ContactUsLinkComponent
 
Glycerine (synthetic) 99+%, pure
 
undefined
Glycerine (synthetic) 99+%, pure
 

 

  • Appearance:
    Clear viscous liquid
  • Infrared spectrum:
    Conforms
  • Refractive index:
    1.4700 to 1.4750 (20°C, 589 nm)
  • GC:
    ≥99.0 %
  • Color scale:
    ≤10 APHA