Order Entry
Finland
Orders LinkContactUsLinkComponent
5-Nonanol ≥98%

5-Nonanol ≥98%

Glass bottle for solids, 25 g
Catalog # L05812.14
5-Nonanol ≥98%
Catalog # L05812.14
CAS Number:  623-93-8
5-Nonanol ≥98%
5-Nonanol ≥98%

Glass bottle for solids, 25 g

CAS Number:  623-93-8
Price
96,60 €
Log in for availability
1 * 25 g
Product Details & Documents

Purity: 98% CAS No.: 623-93-8 Molecular Formula: C9H20O Formula Weight: 144.26 Flash Point: 96°(204°F) Refractive Index: 1.4290 Melting Point: 5° Boiling Point: 194-196° Beilstein Reference: 1733552 MDL No.: MFCD00021944

  • CAS Number:623-93-8
  • Formula:C₉H₂₀O
  • MDL Number:MFCD00021944
  • Molecular Weight:144.26 g/mol
  • Boiling Pt:194…196 °C
  • Melting Pt:5 °C
  • Density:0.826
  • Flash Pt:96 °C (204 °F)
  • Storage Temperature:Ambient
  • EINECS:210-820-2
Specifications

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    25 g

Specifications