Order Entry
Finland
ContactUsLinkComponent
2-Propanol ≥99.7%, AnalaR NORMAPUR® ACS, Reag. Ph. Eur. analytical reagent
2-Propanol ≥99.7%, AnalaR NORMAPUR® ACS, Reag. Ph. Eur. analytical reagent
Supplier:  VWR Chemicals
   Sustainable Options Available
Certificates
undefined
2-Propanol ≥99.7%, AnalaR NORMAPUR® ACS, Reag. Ph. Eur. analytical reagent
Supplier:  VWR Chemicals
   Sustainable Options Available
Certificates

Specifications

  • Environmentally Preferable:
    Safer Human and Environmental Health
  • Environmental certification:
    Chem21 Solvent Selection Guide recommended by default and after discussion

Specifications

Specification Test Results

  • Assay (on anhydrous substance):
    Min. 99.7 %
  • Benzene and related substances:
    Passes test
  • IR Spectrum:
    Passes test
  • Peroxides:
    Passes test
  • Solubility in water:
    Passes test ACS
  • Acidity:
    Max. 0.0001 meq/g
  • Alkalinity:
    Max. 0.0001 meq/g
  • Boiling point:
    81.5 - 82.6 °C
  • Colouration:
    Max. 10 APHA
  • Density (20/4):
    0.784 - 0.786
  • Density (20/20):
    0.785 - 0.787
  • Density (25/25):
    0.783 - 0.790
  • Refractive index (20°C):
    1.376 - 1.378
  • Substances coloured by H2SO4:
    Max. 60 APHA
  • Aldehydes (as CH3CHO):
    Max. 2 ppm
  • Carbonyl compounds (as acetone):
    Max. 20 ppm
  • Carbonyl compounds (as propionaldehyde):
    Max. 20 ppm
  • Ethanol:
    Max. 100 ppm
  • Evaporation residue:
    Max. 10 ppm
  • Formaldehyde:
    Max. 2 ppm
  • Methanol:
    Max. 100 ppm
  • Substances reducing KMnO4 (as O):
    Max. 5 ppm
  • Water:
    Max. 0.1 %
  • Al (Aluminium):
    Max. 0.1 ppm
  • B (Boron):
    Max. 0.02 ppm
  • Ba (Barium):
    Max. 0.05 ppm
  • Ca (Calcium):
    Max. 0.2 ppm
  • Cd (Cadmium):
    Max. 0.01 ppm
  • Co (Cobalt):
    Max. 0.01 ppm
  • Cr (Chromium):
    Max. 0.01 ppm
  • Cu (Copper):
    Max. 0.01 ppm
  • Fe (Iron):
    Max. 0.05 ppm
  • K (Potassium):
    Max. 0.1 ppm
  • Mg (Magnesium):
    Max. 0.05 ppm
  • Mn (Manganese):
    Max. 0.01 ppm
  • Na (Sodium):
    Max. 0.2 ppm
  • Ni (Nickel):
    Max. 0.01 ppm
  • Pb (Lead):
    Max. 0.01 ppm
  • Sn (Tin):
    Max. 0.05 ppm
  • Sr (Strontium):
    Max. 0.05 ppm
  • Zn (Zinc):
    Max. 0.01 ppm
  • Conforms to ACS:
    Passes test
  • Conforms to Reag. Ph.Eur.:
    Passes test

Specification Test Results