1-Nonanol ≥99%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:143-08-8
:
Formula:H₃C(CH₂)₇CH₂OH
MDL Number:MFCD00002990
:144.26 g/mol
Merck Index:14,6679
Boiling Pt:214…216 °C
Melting Pt:–7 °C
Density:0.827
Flash Pt:75 °C (167 °F)
Storage Temperature:Ambient
EINECS:205-583-7
UN:3082
ADR:9,III
:GHS07