1-Nonanol 97%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:143-08-8
:
Formula:H₃C(CH₂)₇CH₂OH
MDL Number:MFCD00002990
:144.26 g/mol
Merck Index:12,06774
Boiling Pt:213…215 °C (760 mmHg)
Melting Pt:–5 °C
Density:0.827 g/cm³
Flash Pt:99 °C
Storage Temperature:Ambient
EINECS:205-583-7
ADR:9,III
:GHS07
Specification Test Results
- Appearance:Clear colorless Liquid
- Infrared spectrum:Conforms
- Refractive index:1.4324 to 1.4344 (20°C, 589 nm)
- GC:≥96.0 %