Order Entry
Finland
Orders LinkContactUsLinkComponent
Diethyl ether

Diethyl ether

Flammable
Irritant
Marketsource
Supplier:   Roth Carl
Diethyl ether
Supplier:  Roth Carl
Marketsource
Certificates
Diethyl ether

2 Options Available Below

Product Details & Documents
Synonyms:  Ethyl Ether, Ether, 2,2'-Oxydiethane, 2,2'-Oxybisethane, Ethoxyethane
  • CAS Number:60-29-7
  • Formula:(CH₃CH₂)₂O
  • MDL Number:MFCD00011646
  • Molecular Weight:74.12 g/mol
  • Merck Index:12,03852
  • Boiling Pt:35 °C (1013 hPa)
  • Melting Pt:–123 °C
  • Density:0.7135 g/cm³ (20 °C)
  • Flash Pt:–40 °C
  • Storage Temperature:Ambient
  • EINECS:200-467-2
  • UN:1155
  • ADR:3,I
  • REACH:01-2119535785-29
Product Family Options

Product Information

  • Pack type
    Pk
    Availability
    Price
  • Glass bottle
    2 L
    Log in for availability
    Log in for availability
    1 * 2 L
    1 * 2 L

    Specifications:
    Marketsource
    Marketsource
  • 35 L
    Log in for availability
    Log in for availability
    1 * 35 L
    1 * 35 L

    Specifications:
    Marketsource
    Marketsource