Nitromethane for synthesis, Sigma-Aldrich®
Catalog # 8.20894.9200
Supplier: Merck
CAS Number:
Specifications
- Pk:200 kg
- Seq:4676
Specifications
Specification Test Results
- Solubility:105 g/l (20 °C)
- Melting Point:-29 °C
- Molar Mass:61.04 g/mol
- Boiling Point:101.2 °C (1013 hPa)
- Vapor Pressure:36.4 hPa (20 °C)
- Flash Point:35.6 °C
- Refractive Index:1.38056 (22 °C, 589 nm)
- Explosion Limit:7.3 - 63.0 %(V)
- Density:1.14 g/cm3 (20 °C)
- pH:6.4 (0.6 g/l, H2O, 20 °C)
- Ignition Point:418 °C
- Saturation concentration:90 g/m3 (20 °C) Air
- Hazard Class:3
- Hazard Class Description:Flammable Liquid
Specification Test Results
About this item
:
:61.04 g/mol
Merck Index:13,06644
Boiling Pt:101.2 °C (1013 hPa)
Melting Pt:–29 °C
Density:1.138 g/cm³ (20 °C)
Flash Pt:44 °C
Storage Temperature:Ambient
EINECS:200-876-6
UN:1261
ADR:3,II
:M-Clarity™ quality level = MQ200
:GHS02
:GHS08
:GHS07
CAS Number:75-52-5
Formula:CH₃NO₂
MDL Number:MFCD00007400