Nitromethane ≥98%
Supplier: Thermo Fisher Scientific
Jump to:
About this item
Purity: 98+% CAS No.: 75-52-5 Molecular Formula: CH3NO2 Formula Weight: 61.04 Flash Point: 35°(95°F) Physical Form: Liquid Density: 1.130 Refractive Index: 1.3820 Appearance: Colourless Odour: Characteristic Melting Point: -29° Boiling Point: 101° Beilstein Reference: 1698205 Merck Reference: 14,6611 UN No.: 1261 MDL No.: MFCD00007400
- CAS Number:75-52-5
- Formula:CH₃NO₂
- MDL Number:MFCD00007400
- Molecular Weight:61.04 g/mol
- Merck Index:14,6611
- Boiling Pt:101 °C
- Melting Pt:–29 °C
- Density:1.130
- Flash Pt:35 °C (95 °F)
- Storage Temperature:Ambient
- EINECS:200-876-6
- UN:1261
- ADR:3,II
2 Options Available Below
Product Details & Documents
Purity: 98+% CAS No.: 75-52-5 Molecular Formula: CH3NO2 Formula Weight: 61.04 Flash Point: 35°(95°F) Physical Form: Liquid Density: 1.130 Refractive Index: 1.3820 Appearance: Colourless Odour: Characteristic Melting Point: -29° Boiling Point: 101° Beilstein Reference: 1698205 Merck Reference: 14,6611 UN No.: 1261 MDL No.: MFCD00007400
- CAS Number:75-52-5
- Formula:CH₃NO₂
- MDL Number:MFCD00007400
- Molecular Weight:61.04 g/mol
- Merck Index:14,6611
- Boiling Pt:101 °C
- Melting Pt:–29 °C
- Density:1.130
- Flash Pt:35 °C (95 °F)
- Storage Temperature:Ambient
- EINECS:200-876-6
- UN:1261
- ADR:3,II



