Order Entry
Finland
Orders LinkContactUsLinkComponent
 

Nitromethane 96% ACS

Flammable
Irritant
Health
Nitromethane 96% ACS
Certificates
Nitromethane 96% ACS

3 Options Available Below

Product Details & Documents
  • CAS Number:75-52-5
  • Formula:CH₃NO₂
  • MDL Number:MFCD00007400
  • Molecular Weight:61.04 g/mol
  • Merck Index:13,06644
  • Boiling Pt:101.2 °C (1013 hPa)
  • Melting Pt:–29 °C
  • Density:1.12 g/cm³
  • Flash Pt:35 °C
  • Storage Temperature:Ambient
  • EINECS:200-876-6
  • UN:1261
  • ADR:3,II

 

Specifications for Product Family

Specification Test Results

  • Appearance:
    Clear liquid
  • Infrared spectrum:
    Conforms
  • Refractive index:
    1.3805 to 1.3825 (20°C, 589 nm)
  • Water:
    ≤0.05 % (Coulometric)
  • GC:
    ≥95 %
  • Color scale:
    ≤10 APHA
Product Family Options

Product Information

  • Pack type
    Pk
    Saatavuus
    Price
  • Glass bottle
    500 mL
    Log in for availability
    Log in for availability
    1 * 500 mL
    1 * 500 mL - 107,00 €
    107,00 €
    1 * 500 mL
       107,00 €

  • Glass bottle
    1 L
    Log in for availability
    Log in for availability
    1 * 1 L
    1 * 1 L - 174,00 €
    174,00 €
    1 * 1 L
       174,00 €

  • Glass bottle
    2,5 L
    Log in for availability
    Log in for availability
    1 * 2,5 L
    1 * 2,5 L - 318,00 €
    318,00 €
    1 * 2,5 L
       318,00 €