1-Iodoheptane ≥98% stabilised
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:4282-40-0
:
Formula:C₇H₁₅I
MDL Number:MFCD00001104
:226.1 g/mol
Boiling Pt:203…205 °C
Melting Pt:–48 °C
Density:1.379
Flash Pt:78 °C (174 °F)
:Stabilised with copper
EINECS:224-285-8
:GHS07
Storage Temperature:Ambient