0,00 €
Order Entry
/
Products
Services
Digital Solutions
Applications
Support
About
Finland
ContactUsLinkComponent
1-Chlorooctane
1-Chlorooctane ≥99%
1-Chlorooctane ≥99%
:
Thermo Fisher Scientific
1-Chlorooctane ≥99%
:
Thermo Fisher Scientific
:
111-85-3
:
:
C₈H₁₇Cl
:
MFCD00001022
:
148.68 g/mol
:
182…183 °C
:
–58 °C
:
0.873
:
63 °C (145 °F)
:
Ambient
:
203-915-5
:
3082
:
9,III
:
GHS09
msds_ALFAL04358
Organic Building Blocks and Intermediates
Thermo Scientific Acroseal 25 Years