


Thallium(I) acetate ≥99.995% (metals basis)
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:563-68-8
:
Formula:C₂H₃O₂Tl
MDL Number:MFCD00013045
:263.43 g/mol
Merck Index:14,9257
Boiling Pt:1457 °C (1013 hPa)
Melting Pt:131 °C
Density:3.68 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:209-257-5
UN:1707
ADR:6.1,II
:GHS06
:GHS08
:GHS09