Order Entry
Finland
Orders LinkContactUsLinkComponent
 

Behenic acid

Glass bottle for solids, 1 kg
Catalog # ROTH5030.1
Marketsource
Supplier:   Roth Carl
Behenic acid
Catalog # ROTH5030.1
Supplier:  Roth Carl
Marketsource
CAS Number:  112-85-6
Behenic acid

Glass bottle for solids, 1 kg

CAS Number:  112-85-6
Price
Inquire For Price
Log in for availability
1 * 1 kg
Product Details & Documents
Synonyms:  Docosanoic acid
  • CAS Number:112-85-6
  • Formula:C₂₂H₄₄O₂
  • MDL Number:MFCD00002807
  • Molecular Weight:340.59 g/mol
  • Merck Index:13,01024
  • Boiling Pt:306 °C (1013 hPa)
  • Melting Pt:80 °C
  • Density:0.8221 g/cm³ (20 °C)
  • Storage Temperature:Ambient
  • EINECS:204-010-8
Specifications

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    1 kg

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.