Order Entry
Finland
Orders LinkContactUsLinkComponent
Acetylacetone

Acetylacetone

Glass bottle for solids, 1 kg
Catalog # MOLEM78787938
Flammable
Irritant
Marketsource
Supplier:   Molekula
Acetylacetone
Catalog # MOLEM78787938
Supplier:  Molekula
Marketsource
CAS Number:  123-54-6
Acetylacetone
Acetylacetone

Glass bottle for solids, 1 kg

CAS Number:  123-54-6
Price
81,00 €
Log in for availability
1 * 1 kg
Product Details & Documents
Synonyms:  ACAC, 2,4-Pentanedione
  • CAS Number:123-54-6
  • Formula:CH₃COCH₂COCH₃
  • MDL Number:MFCD00008787
  • Molecular Weight:100.12 g/mol
  • Merck Index:13,00083
  • Boiling Pt:140.4 °C (1013 hPa)
  • Melting Pt:–23 °C
  • Density:0.96945 g/cm³ (20 °C)
  • Flash Pt:30 °C
  • Storage Temperature:Ambient
  • EINECS:204-634-0
  • UN:2310
  • ADR:3,III
Specifications

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    1 kg

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.