Acetone ≥99.8%, AnalaR NORMAPUR® ACS, Reag. Ph. Eur. analytical reagent
:67-64-1
:
:CH₃COCH₃
:MFCD00008765
:58.08 g/mol
:13,00067
:56.2 °C (1013 hPa)
:–95.4 °C
:0.792 g/cm³ (20 °C)
:<–20 °C (closed cup)
:Ambient
:200-662-2
:1090
:3,II
:01-2119471330-49
:GHS02
:GHS07
- Environmentally Preferable:Safer Human and Environmental Health
- Environmental certification:Chem21 Solvent Selection Guide recommended after discussion
- Assay (on anhydrous substance):Min. 99.8 %
- Appearance of solution:Passes test Ph.Eur.
- Aqueous solution:Passes test
- Identification B:Passes test Ph.Eur.
- Identification C:Passes test Ph.Eur.
- Insolubility in water:Passes test
- IR Spectrum:Passes test
- Related substances:Passes test Ph.Eur.
- Acidity:Max. 0.0003 meq/g
- Alkalinity:Max. 0.0003 meq/g
- Boiling point:56.0 - 56.7 °C
- Colouration:Max. 10 APHA
- Density (20/4):0.790 - 0.792
- Density (20/20):0.790 - 0.793
- Aldehydes (as HCHO):Max. 10 ppm
- Ethanol:Max. 100 ppm
- Evaporation residue:Max. 5 ppm
- Methanol:Max. 0.05 %
- Propan-2-ol:Max. 100 ppm
- Substances reducing KMnO4 (as O):Max. 2 ppm
- Water:Max. 0.2 %
- PO4 (Phosphate):Max. 100 ppb
- Al (Aluminium):Max. 0.1 ppm
- B (Boron):Max. 0.02 ppm
- Ba (Barium):Max. 0.05 ppm
- Ca (Calcium):Max. 0.5 ppm
- Cd (Cadmium):Max. 0.01 ppm
- Co (Cobalt):Max. 0.01 ppm
- Cr (Chromium):Max. 0.02 ppm
- Cu (Copper):Max. 0.01 ppm
- Fe (Iron):Max. 0.05 ppm
- K (Potassium):Max. 0.1 ppm
- Mg (Magnesium):Max. 0.05 ppm
- Mn (Manganese):Max. 0.01 ppm
- Na (Sodium):Max. 0.5 ppm
- Ni (Nickel):Max. 0.01 ppm
- Pb (Lead):Max. 0.01 ppm
- Sn (Tin):Max. 0.1 ppm
- Sr (Strontium):Max. 0.02 ppm
- Zn (Zinc):Max. 0.01 ppm
- Conforms to ACS:Passes test
- Conforms to Reag. Ph.Eur.:Passes test