Acetone ≥99.5% ACS
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:67-64-1
:
Formula:CH₃COCH₃
MDL Number:MFCD00008765
:58.08 g/mol
Merck Index:14,66
Boiling Pt:56 °C
Melting Pt:–94 °C
Density:0.791
Flash Pt:–17 °C (1 °F)
Storage Temperature:Ambient
EINECS:200-662-2
UN:1090
ADR:3,II
REACH:01-2119471330-49
:GHS02
:GHS07
Specifications
- Environmentally Preferable:Safer Human and Environmental Health
- Environmental certification:Chem21 Solvent Selection Guide recommended after discussion