Acetone 99.6% ACS
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:67-64-1
:
Formula:CH₃COCH₃
MDL Number:MFCD00008765
:58.08 g/mol
Merck Index:13,00067
Boiling Pt:56 °C (760 mmHg)
Melting Pt:–95 °C
Density:0.79 g/cm³
Flash Pt:–20 °C
Storage Temperature:Ambient
EINECS:200-662-2
UN:1090
ADR:3,II
REACH:01-2119471330-49
:GHS07
:GHS02
Specifications
- Environmentally Preferable:Safer Human and Environmental Health
- Environmental certification:Chem21 Solvent Selection Guide recommended after discussion
Specifications
Specification Test Results
- Appearance:Clear liquid
- Water:≤0.5 % (K.F.)
- Solubility:(in water) Passes test
- GC:≥99.5 %
- Color scale:≤10 APHA
- Residue after evaporation:≤0.001 %
- Isopropyl alcohol:≤0.05 %
- Titrable acid:≤0.0003 meq/g
- Titrable base:≤0.0006 meq/g
- Aldehyde (HCHO):≤0.002 %
- Methanol:≤0.05 %
- KMnO4-reducing substance:Passes test