

4-Heptanone
Catalog # PROOCHE150
Supplier: LGC Standards PROMOCHEM
CAS Number:
Specifications
- Pack type:Glass bottle
- Pk:2 mL
- Seq:2145
Specifications
About this item
:
:114.19 g/mol
Merck Index:13,03379
Boiling Pt:144 °C (1013 hPa)
Melting Pt:–34 °C
Density:0.8116 g/cm³ (20 °C)
Flash Pt:48 °C
Storage Temperature:Ambient
EINECS:204-608-9
UN:2710
ADR:3,III
:GHS02
:GHS07
CAS Number:123-19-3
Formula:C₇H₁₄O
MDL Number:MFCD00009403