Order Entry
Finland
ContactUsLinkComponent
3-Heptanone
Flammable
Irritant
3-Heptanone
Catalog # PROOCHE149
CAS Number:  
Flammable
Irritant
3-Heptanone
Catalog # PROOCHE149
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    2 mL
  • Seq:
    2145

Specifications

About this item

 :
 :114.19 g/mol
Boiling Pt:147 °C (1013 hPa)
Melting Pt:–37 °C
Density:0.812 g/cm³ (20 °C)
Flash Pt:41 °C
Storage Temperature:Ambient
EINECS:203-388-1
UN:1224
ADR:3,III
 :GHS02
 :GHS07
CAS Number:106-35-4
Formula:C₇H₁₄O
MDL Number:MFCD00009483