Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Finland
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Cytochalasin B
Cytochalasin B
Cytochalasin B
A7657.0005
:
PanReac AppliChem
:
Cytochalasin B
A7657.0005
:
PanReac AppliChem
:
Pack type:
Glass bottle for solids
Pk:
5 mg
Seq:
2210
:
:
479.62 g/mol
:
218…221 °C
:
239-000-2
:
1544
:
6.1,II
:
14930-96-2
:
C₂₉H₃₇NO₅
:
MFCD00077704