
5-Bromo-1-pentanol ≥90.0% (by GC)
Supplier: TCI
Certificates
About this item
CAS Number:34626-51-2
:
Formula:C₅H₁₁BrO
MDL Number:MFCD00061101
:167.05 g/mol
Boiling Pt:117°C
:GHS07
Storage Temperature:Freezer
Specification Test Results
- Purity (GC):min. 90.0 %