Order Entry
Finland
ContactUsLinkComponent
 
 
4-Quinolinecarbaldehyde ≥97.0% (by GC, titration analysis)
 :  TCI
 
undefined
 
4-Quinolinecarbaldehyde ≥97.0% (by GC, titration analysis)
 :  TCI

 

  • Appearance:
    White to Yellow to Orange powder to crystal
  • Purity (GC):
    min. 97.0 %
  • Purity (Nonaqueous Titration):
    min. 97.0 %
  • Melting point:
    47.0 to 53.0 deg-C