
5-Bromo-2-methoxybenzenesulphonyl chloride ≥98%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:23095-05-8
:
Formula:C₇H₆BrClO₃S
MDL Number:MFCD00051768
:285.55 g/mol
Melting Pt:112…115 °C
Storage Temperature:Ambient
UN:3261
ADR:8,II
:GHS05