
2-Chloro-1-methylpyridinium iodide 97%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:14338-32-0
:
Formula:C₆H₇ClIN
MDL Number:MFCD00011984
:255.49 g/mol
Melting Pt:195…200 °C
EINECS:238-288-7
:GHS07
Specification Test Results
- Appearance:Yellow Crystalline powder and/or chunks
- Infrared spectrum:Conforms
- Melting point:195 to 200 °C
- Titration Argentometric:96.0 to 104.0 %
- Solubility:light yellow
- HPLC:≥97.5 %