Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Finland
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
2-Bromo-5-(trifluoromethoxy)phenol
2-Bromo-5-(trifluoromethoxy)phenol ≥97%
2-Bromo-5-(trifluoromethoxy)phenol ≥97%
Toxic
Irritant
2-Bromo-5-(trifluoromethoxy)phenol ≥97%
Supplier:
Thermo Fisher Scientific
Certificates
Expand image
Toxic
Irritant
2-Bromo-5-(trifluoromethoxy)phenol ≥97%
Supplier:
Thermo Fisher Scientific
Certificates
About this item
CAS Number:
205371-26-2
:
Formula:
C₇H₄BrF₃O₂
MDL Number:
MFCD06660162
:
257.01 g/mol
EINECS:
None
:
GHS06
:
GHS07
Storage Temperature:
Ambient
Related Information
Documentation
Certificates
Search by Lot Number
msds_ALFAH26389
Application Notes
Organic Building Blocks and Intermediates
Videos
Thermo Scientific Acroseal 25 Years
Documentation