2,4-Dinitro-1-naphthol, dark yellow powder C.I. 10315
Supplier: MP Biomedicals
Certificates
About this item
CAS Number:605-69-6
:
Formula:C₁₀H₆N₂O₅
MDL Number:MFCD00065352
:234.17 g/mol
Boiling Pt:408 °C (1013 hPa)
Melting Pt:130…132 °C (dec.)
Storage Temperature:Ambient
EINECS:210-093-1
:GHS07
Specification Test Results
- Identity Test:Passes