2,4-Dichlorophenol
Catalog # ACCUA-017
Supplier: AccuStandard
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:100 mg
- Seq:2440
Specifications
About this item
:
:163 g/mol
Merck Index:13,03098
Boiling Pt:210 °C (1013 hPa)
Melting Pt:40…41 °C
Density:1.4 g/cm³ (20 °C)
Flash Pt:114 °C
EINECS:204-429-6
UN:2928
ADR:6.1,II
:GHS06
:GHS05
:GHS09
Storage Temperature:Ambient
CAS Number:120-83-2
Formula:C₆H₄Cl₂O
MDL Number:MFCD00002169