Order Entry
Finland
ContactUsLinkComponent
Supelco® Organic Reference Standard, Heptacosafluorotributylamine
Supelco® Organic Reference Standard, Heptacosafluorotributylamine
Supplier:  Merck
Certificates
undefined
Supelco® Organic Reference Standard, Heptacosafluorotributylamine
Supplier:  Merck

Specifications

  • CAS No.:
    311-89-7
  • Storage conditions:
    RT
  • Shipping conditions:
    STD
  • MDL No.:
    MFCD00000436
  • Synonym(s):
    PFTBA; Perfluorotributylamine; Tris(nonafluorobutyl)amine
  • Formula:
    N(CF₂CF₂CF₂CF₃)₃
  • Molecular weight:
    671,09
  • Density:
    1,883 g/ml at 25 °C (lit.)
  • Boiling point:
    178 °C (lit.)

Specifications