Lithium hydroxide monohydrate
Marketsource
Supplier: Roth Carl
Jump to:
About this item
- CAS Number:1310-66-3
- Formula:LiOH·1H₂O
- MDL Number:MFCD00149772
- Molecular Weight:41.96 g/mol
- Merck Index:13,05556
- Boiling Pt:920 °C (1013 hPa)
- Melting Pt:462 °C
- Density:1.51 g/cm³ (20 °C)
- Storage Temperature:Ambient
- EINECS:215-183-4
- UN:2680
- ADR:8,II
2 Options Available Below
Product Details & Documents
- CAS Number:1310-66-3
- Formula:LiOH·1H₂O
- MDL Number:MFCD00149772
- Molecular Weight:41.96 g/mol
- Merck Index:13,05556
- Boiling Pt:920 °C (1013 hPa)
- Melting Pt:462 °C
- Density:1.51 g/cm³ (20 °C)
- Storage Temperature:Ambient
- EINECS:215-183-4
- UN:2680
- ADR:8,II
Product Family Options
Product Information
- Pack typePkAvailabilityPrice
- Specifications:MarketsourceMarketsource
- Specifications:MarketsourceMarketsource



