Lithium hydroxide monohydrate
Glass bottle for solids, 1 kg
Catalog # MOLEM49289225
Marketsource
Supplier: Molekula
Lithium hydroxide monohydrate
Catalog # MOLEM49289225
Supplier: Molekula
Marketsource
CAS Number: 1310-66-3
Jump to:
Product Details & Documents
- CAS Number:1310-66-3
- Formula:LiOH·1H₂O
- MDL Number:MFCD00149772
- Molecular Weight:41.96 g/mol
- Merck Index:13,05556
- Boiling Pt:920 °C (1013 hPa)
- Melting Pt:462 °C
- Density:1.51 g/cm³ (20 °C)
- Storage Temperature:Ambient
- EINECS:215-183-4
- UN:2680
- ADR:8,II
Specifications
Specifications
- Pack type:Glass bottle for solids
- Pk:1 kg
- Cat. No.:MOLEM49289225
Specifications
Suositukset
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



