Order Entry
Finland
ContactUsLinkComponent
di-Sodium tetraborate decahydrate ≥98%, purified
Health
di-Sodium tetraborate decahydrate ≥98%, purified
Catalog # 27721.366
Supplier:  VWR Chemicals
CAS Number:  
undefined
Health
di-Sodium tetraborate decahydrate ≥98%, purified
Catalog # 27721.366
Supplier:  VWR Chemicals
CAS Number:  

Specifications

  • Pack type:
    Plastic bucket
  • Pk:
    5 kg
  • Seq:
    4114

Specifications

Specification Test Results

  • Assay:
    Min. 98 %
  • Heavy metals (as Pb):
    Max. 20 ppm
  • Fe (Iron):
    Max. 20 ppm

Specification Test Results

About this item

 :
 :381.37 g/mol
Merck Index:13,08662
Boiling Pt:320 °C (1013 hPa)
Melting Pt:75 °C
Density:1.73 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:215-540-4
REACH:01-2119490790-32
 :GHS08
CAS Number:1303-96-4
Formula:Na₂B₄O₇·10H₂O
MDL Number:MFCD00149193