Order Entry
Finland
ContactUsLinkComponent
Hydrochloric acid 0.04 mol/l (0.04 N), AVS TITRINORM according to the NF T 90-036 standard, volumetric solution
Corrosive
Hydrochloric acid 0.04 mol/l (0.04 N), AVS TITRINORM according to the NF T 90-036 standard, volumetric solution
Catalog # 30022.293
Supplier:  VWR Chemicals
CAS Number:  
undefined
Corrosive
Hydrochloric acid 0.04 mol/l (0.04 N), AVS TITRINORM according to the NF T 90-036 standard, volumetric solution
Catalog # 30022.293
Supplier:  VWR Chemicals
CAS Number:  

Specifications

  • Pack type:
    Plastic bottle
  • Pk:
    1 L
  • Seq:
    5037

Specifications

Specification Test Results

  • NIST traceable:
    Confirmed
  • Titer (20°C; real value 0.2 % accuracy):
    0.03992 - 0.04008 mol/l

Specification Test Results

About this item

 :
Density:1.004 g/cm³ (25 °C)
Storage Temperature:Ambient
EINECS:231-595-7
ADR:8,III
 :36.46 g/mol
UN:1789
 :GHS05
CAS Number:7647-01-0
Formula:HCl
MDL Number:MFCD00011324