α-Pinene 97% stabilised
Supplier: Thermo Fisher Scientific
About this item
- CAS Number:80-56-8
- Formula:C₁₀H₁₆
- MDL Number:MFCD00001339
- Molecular Weight:136.24 g/mol
- Boiling Pt:154…157 °C (760 mmHg)
- Melting Pt:–55 °C
- Density:0.85 g/cm³
- Flash Pt:38 °C
- Storage Temperature:Ambient
- EINECS:219-445-9
- UN:2368
- ADR:3,III
- :Stabilised with α-tocopherol
- :Density: 0.8592 g/cm³ (20 °C)
3 Options Available Below
Product Details & Documents
- CAS Number:80-56-8
- Formula:C₁₀H₁₆
- MDL Number:MFCD00001339
- Molecular Weight:136.24 g/mol
- Boiling Pt:154…157 °C (760 mmHg)
- Melting Pt:–55 °C
- Density:0.85 g/cm³
- Flash Pt:38 °C
- Storage Temperature:Ambient
- EINECS:219-445-9
- UN:2368
- ADR:3,III
- :Stabilised with α-tocopherol
- :Density: 0.8592 g/cm³ (20 °C)
Specifications for Product Family
Specification Test Results
- Appearance:Clear colorless Liquid
- Infrared spectrum:Conforms
- Refractive index:1.4640 to 1.4680 (20°C, 589 nm)
- GC:≥96.0 %
- Stabilizer:0.8 % Alpha-tocopherol



