Order Entry
Finland
ContactUsLinkComponent
(±)-2-Butanol, EMSURE® for analysis, Supelco®
Flammable
Irritant
(±)-2-Butanol, EMSURE® for analysis, Supelco®
Supplier:  Merck
Certificates
undefined
Flammable
Irritant
(±)-2-Butanol, EMSURE® for analysis, Supelco®
Supplier:  Merck
Certificates

Specification Test Results

  • Solubility:
    125 g/l (20 °C)
  • Melting Point:
    -115 °C
  • Molar Mass:
    74.12 g/mol
  • Boiling Point:
    99 - 102 °C (1013 hPa)
  • Vapor Pressure:
    16.5 hPa (20 °C)
  • Flash Point:
    24 °C
  • Explosion Limit:
    1.4 - 9.8 %(V)
  • Density:
    0.81 g/cm3 (20 °C)
  • pH:
    (H2O, 20 °C) neutral
  • Ignition Point:
    390 °C
  • Evaporation number:
    20

Specification Test Results

Related Information

Certificates
Search by Lot Number