Order Entry
Finland
ContactUsLinkComponent
 
 
Orthophosphoric acid 85% ≥85%, TECHNICAL
  90045.5000
 :  
undefined
 
Orthophosphoric acid 85% ≥85%, TECHNICAL
  90045.5000
 :  

 

  • Pack type:
    Plastic container
  • Pk:
    5 L
  • Seq:
    5190

 

 

  • Assay:
    Min. 85 %

 

 

 :
 :98 g/mol
 :28 °C
 :1.689 g/cm³ (20 °C)
 :Ambient
 :231-633-2
 :1805
 :8,III
 :GHS05
 :7664-38-2
 :H₃PO₄
 :MFCD00011340