Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Finland
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Sodium hydroxide
Sodium hydroxide 50% in aqueous solution, TECHNICAL
Sodium hydroxide 50% in aqueous solution, TECHNICAL
:
VWR Chemicals
Sodium hydroxide 50% in aqueous solution, TECHNICAL
:
VWR Chemicals
:
1310-73-2
:
:
NaOH
:
MFCD00003548
:
40 g/mol
:
119…135 °C (1013 hPa)
:
9…10 °C
:
1.53 g/cm³ (20 °C)
:
Ambient
:
215-185-5
:
1824
:
8,II
:
GHS05
Assay:
47 - 51 %
2,5L Plastic bottle for liquids DIN 45 UN X (ZPACK13962500)
25L Plastic drum for liquids DIN 61 (ZPACK13952500)
5L Plastic bottle for liquids DIN 45 UN X (ZPACK13951035)
SDS_EN_UK_VWRC28265.pdf
sds_VWRC28265
spec_VWRC28265.322
spec_VWRC28265.366
spec_VWRC28265.446
VWR Chemicals Big Packs and Bulks